Merck
Formulation and drying of miconazole and itraconazole nanosuspensions.
International journal of pharmaceutics (2013-01-08)
Ana M Cerdeira, Marco Mazzotti, Bruno Gander
ZUSAMMENFASSUNG
Miconazole and itraconazole possess adequate membrane permeability, but only slight water solubility, which limits their bioavailability and antifungal effect. To increase their dissolution rate, the compounds were nanoground by media milling to produce nanosuspensions with mean particle size of approximately 210 nm and stabilized with sodium dodecylsulfate (SDS) in combinations with either cellulose ethers (HPC or HPMC) or poloxamers. During storage for 3 months at 25 °C, HPC/SDS stabilized more efficiently miconazole nanoparticles, while poloxamer 407/SDS performed better with itraconazole nanosuspensions. The stabilizing efficiency of the excipients was explained by physical-chemical drug-excipients interactions. The HPC/SDS-stabilized nanosuspensions were spray-dried or freeze-dried with and without the matrix formers mannitol or microcrystalline cellulose (MCC). In absence of matrix former, itraconazole particles agglomerated more extensively than miconazole particles, resulting in a low dissolution rate. Dissolution of the spray- or freeze-dried miconazole nanosuspension was enhanced in presence of mannitol or MCC (drug substance:excipient ratio of 1:1, w/w), as compared to the coarse drug suspension (twice the amount dissolved after 10 and 20 min). Spray-drying itraconazole nanosuspension in presence of mannitol or MCC also yielded fast dissolution (60% dissolved in less than 10 min as compared to 30-45 min with the coarse suspension). Freeze-dried itraconazole nanosuspensions did generally not dissolve substantially faster than freeze-dried coarse suspension. In conclusion, we were able to process miconazole and itraconazole successfully and under similar conditions into dry nanoparticulate drug products with enhanced in vitro performance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methylzellulose, viscosity: 4,000 cP
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methylzellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydroxypropylcellulose, average Mw ~80,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Hydroxypropylcellulose, average Mw ~100,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~86,000
Sigma-Aldrich
Methylzellulose, viscosity: 1,500 cP
Sigma-Aldrich
(Hydroxypropyl)methylcellulose
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hydroxypropylcellulose, average Mw ~370,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Methylzellulose, viscosity: 15 cP
Sigma-Aldrich
Methylzellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
Methylzellulose, viscosity: 25 cP
Sigma-Aldrich
Methylzellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methylzellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~120,000
Sigma-Aldrich
Methocel® A15 LV, 27.5-31.5% methoxyl basis
Sigma-Aldrich
(±)-Miconazol -nitrat (Salz), imidazole antibiotic
Sigma-Aldrich
Hydroxypropylcellulose, average Mw ~1,000,000, powder, 20 mesh particle size (99% through)
Supelco
(±)-Miconazol -nitrat (Salz), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~90,000
Sigma-Aldrich
Methocel® MC, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methylzellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methylzellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Miconazolnitrat, European Pharmacopoeia (EP) Reference Standard
Miconazol, European Pharmacopoeia (EP) Reference Standard