Merck
  • [Dependence of the radioprotective effect of beta-mercaptoethylamine on the thiol level of the outer cell membrane].
[Dependence of the radioprotective effect of beta-mercaptoethylamine on the thiol level of the outer cell membrane].
Radiobiologiia (1989-03-01)
M M Konstantinova, G V Dontsova, N M Koroleva
ZUSAMMENFASSUNG
In experiments with Ehrlich ascites tumor cells with the use of a specific thiol blocker, 6,6'-dithiodinicotinic acid, it was shown that thiols of a cell plasmic membrane are necessary for the radioprotective effect of mercaptoethylamine to be realized.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6,6′-Dithiodinikotinsäure, technical grade, 85%