Merck
  • Constriction to ETB receptor agonists, BQ-3020 and sarafotoxin s6c, in human resistance and capacitance vessels in vivo.
Constriction to ETB receptor agonists, BQ-3020 and sarafotoxin s6c, in human resistance and capacitance vessels in vivo.
British journal of clinical pharmacology (2000-07-25)
F E Strachan, T R Crockett, N L Mills, G A Gray, D J Webb
ZUSAMMENFASSUNG
The aim of the study was to examine the effects of the ETB receptor selective agonists sarafotoxin S6c (SFTX6c) and BQ-3020 on the forearm resistance and capacitance vessels in healthy subjects in vivo. The local response to intra-arterial or intravenous infusion of SFTX6c (5 pmol min-1) or BQ-3020 (50 pmol min-1) was assessed, on separate occasions, in eight healthy men (aged 20-28 years). Data (mean +/- s.e.mean) were examined by ANOVA. Results are expressed as percentage change from baseline at 90 min. SFTX6c and BQ-3020 reduced forearm blood flow, following local intra-arterial infusion (-25 +/- 7% and -27 +/- 7%, respectively; P < 0.001) and reduced hand vein diameter, following local intravenous infusion (-30 +/- 8% and -16 +/- 7%, respectively; P < 0.001). We have shown that locally active infusions of the selective ETB receptor agonists SFTX6c and BQ-3020 cause arterial constriction and venoconstriction in healthy human blood vessels in vivo. These results indicate that ETB receptor stimulation may mediate vasoconstriction in humans.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
BQ-3020