Merck
  • Validated quantitative structure-activity relationship analysis of a series of 2-aminothiazole based p56(Lck) inhibitors.
Validated quantitative structure-activity relationship analysis of a series of 2-aminothiazole based p56(Lck) inhibitors.
Analytica chimica acta (2008-12-03)
Jiazhong Li, Juan Du, Lili Xi, Huanxiang Liu, Xiaojun Yao, Mancang Liu
ZUSAMMENFASSUNG
In this study, the quantitative structure-activity relationship (QSAR) of a series of 2-aminothiazole based Lck inhibitors was investigated. The key structural features responsible for the inhibition activities were discussed in detail. A population of 100 rigorously validated linear QSAR models were established based on the descriptors calculated in DRAGON program and selected by genetic algorithm (GA). A total of 36 descriptors were involved in all the QSAR models. Then the common descriptors appeared in all the models were extracted to build the final QSAR model. As a result, the final 8-parameter QSAR model was established. After analysis of the eight descriptors, some advice was proposed to help the design of possible novel inhibitors with higher bioactivity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Aminothiazol, 97%