Merck
Improvement of chloroperoxidase stability by covalent immobilization on chitosan membranes.
Biotechnology letters (2009-05-01)
Li-Hua Zhang, Chao-Hong Bai, Ying-Song Wang, Yu-Cheng Jiang, Man-Cheng Hu, Shu-Ni Li, Quan-Guo Zhai
ZUSAMMENFASSUNG
Chloroperoxidase (CPO) from Caldariomyces fumago was optimally covalently immobilized on chitosan membranes pretreated with 0.8 M glutaraldehyde at pH 3.5 to give 3.18 mg CPO g(-1) support. Using monochlorodimedone (MCD) as assay substrate, the immobilized-CPO retained 40% activity at 50 degrees C after 40 min whereas free CPO retained only 0.02%. The residual activity for immobilized-CPO was 99 and 58% compared with 68 and 43% for free CPO in the presence of 1.5 M urea and 300 microM H(2)O(2), respectively, after 20 h.