Merck
Assessing the drinking status of liver transplant patients with alcoholic liver disease.
Liver transplantation : official publication of the American Association for the Study of Liver Diseases and the International Liver Transplantation Society (2013-01-03)
John P Allen, Friedrich M Wurst, Natasha Thon, Raye Z Litten
ZUSAMMENFASSUNG
The accurate assessment of drinking by patients with alcoholic liver disease is important both before and after liver transplantation. Unfortunately, self-reports by these individuals often underestimate their actual alcohol consumption. Several recently developed biochemical measures can provide additional information on a patient's use of alcohol. This article describes ethyl glucuronide, ethyl sulfate, phosphatidyl ethanol, and carbohydrate-deficient transferrin as biomarkers of drinking and summarizes research dealing with their application in patients with alcohol use disorders who are candidates for or recipients of liver transplantation. The article also offers suggestions for enhancing the reliability of self-report measures of drinking status.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Ethyl-β-D-glucuronid, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Diethylsulfat, 98%
Supelco
Ethyl-β-D-glucuronid, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®