Merck
  • Kinetic studies of urokinase-catalysed hydrolysis of 5-oxo-L-prolylglycyl-L-arginine 4-nitroanilide.
Kinetic studies of urokinase-catalysed hydrolysis of 5-oxo-L-prolylglycyl-L-arginine 4-nitroanilide.
Biochimica et biophysica acta (1980-06-13)
H H Ipsen, U Christensen
ZUSAMMENFASSUNG
The enzymic properties of urokinase (EC 3.4.21.31) were studied. The kinetic parameters of hydrolysis of 5-oxo-Pro-Gly-Arg-NA were determined in the pH range 5-9, at 25 degrees C and 37 degrees C. The reaction is affected by only one ionizing group of urokinase with pK 7.15 (25 degrees C) and pK 6.82 (37 degrees C). The results indicate that 5-oxo-Pro-Gly-Arg-NA is a good model substrate for studies of the conversion of plasminogen to plasmin. The Km values of the urokinase-catalysed hydrolysis of plasminogen and 5-oxo-Pro-Gly-Arg-NA are of the same order of magnitude. Plasmin catalyses the hydrolysis of 5-oxo-Pro-Gly-Arg-NA, but the Km value is several hundred times that of urokinase. Urokinase is shown not to react with good plasmin substrates, such as Bz-Arg-OEt and D-Val-Leu-Lys-NA, but is linearly competitively inhibited by 6-amino-hexanoic acid and trans-4-aminomethylcyclohexane-1-carboxylic acid.