Merck
  • Supercooperativity in platelet aggregation: substituted pyridyl isoxazoles, a new class of supercooperative platelet aggregation inhibitors.
Supercooperativity in platelet aggregation: substituted pyridyl isoxazoles, a new class of supercooperative platelet aggregation inhibitors.
FEBS letters (1994-09-05)
P V Vrzheshch, O V Demina, S I Shram, S D Varfolomeev
ZUSAMMENFASSUNG
The phenomenon of supercooperativity in platelet aggregation is manifested by the occurrence of clear-cut thresholds in dose-response relationships; in such cases the Hill coefficient has unusually high values. Approximation, by the Hill equation, of the relationship of the rate of arachidonate-induced platelet aggregation to the concentrations of either the inducer or inhibitors such as substituted pyridyl isoxazoles (synthesized by us), indomethacin, and pinane thromboxane A2, demonstrated that the Hill coefficients ranged from 30 to 100. 3-(3-Pyridyl)-5-phenylisoxazole, which exhibited maximal anti-aggregatory activity among the synthesized compounds, inhibited neither cyclooxygenase nor thromboxane synthase. The compounds affected the signal transduction pathway at/or posterior to the stage of thromboxane A2 reception.