Merck
  • AK2 activates a novel apoptotic pathway through formation of a complex with FADD and caspase-10.
AK2 activates a novel apoptotic pathway through formation of a complex with FADD and caspase-10.
Nature cell biology (2007-10-24)
Ho-June Lee, Jong-Ok Pyo, Yumin Oh, Hyo-Jin Kim, Se-hoon Hong, Young-Jun Jeon, Hyunjoo Kim, Dong-Hyung Cho, Ha-Na Woo, Sungmin Song, Jung-Hyun Nam, Hyo Joon Kim, Key-Sun Kim, Yong-Keun Jung
ZUSAMMENFASSUNG
Mitochondrial proteins function as essential regulators in apoptosis. Here, we show that mitochondrial adenylate kinase 2 (AK2) mediates mitochondrial apoptosis through the formation of an AK2-FADD-caspase-10 (AFAC10) complex. Downregulation of AK2 attenuates etoposide- or staurosporine-induced apoptosis in human cells, but not that induced by tumour-necrosis-factor-related apoptosis-inducing ligand (TRAIL) or Fas ligand (FasL). During intrinsic apoptosis, AK2 translocates to the cytoplasm, whereas this event is diminished in Apaf-1 knockdown cells and prevented by Bcl-2 or Bcl-X(L). Addition of purified AK2 protein to cell extracts first induces activation of caspase-10 via FADD and subsequently caspase-3 activation, but does not affect caspase-8. AFAC10 complexes are detected in cells undergoing intrinsic cell death and AK2 promotes the association of caspase-10 with FADD. In contrast, AFAC10 complexes are not detected in several etoposide-resistant human tumour cell lines. Taken together, these results suggest that, acting in concert with FADD and caspase-10, AK2 mediates a novel intrinsic apoptotic pathway that may be involved in tumorigenesis.