Merck
  • Gas-liquid chromatographic and mass spectrometric indentification of chlorinated trifluorotoluene residues in Niagara River fish.
Gas-liquid chromatographic and mass spectrometric indentification of chlorinated trifluorotoluene residues in Niagara River fish.
Journal - Association of Official Analytical Chemists (1979-01-01)
M P Yurawecz
PMID422503
ZUSAMMENFASSUNG
Several unknown halogenated compounds were detected in Niagara River fish using a method similar to the AOAC multiresidue method for chlorinated pesticides in high-moisture foods. From gas-liquid chromatographic-mass spectrometric (GLC/MS) data and GLC retention times on 3 columns, 7 of the compounds were identified as 4-chloro-alpha,alpha,alpha-trifluorotoluene (0.17--2.0 ppm), 2-chloro-alpha,alpha,alpha-trifluorotoluene (0.002--0.1 ppm), 3,4-dichloro-alpha,alpha,alpha-trifluorotoluene (0.02--0.28 ppm), 2,4-dichloro-alpha,alpha-alpha-trifluorotoluene (0.02--0.17) ppm), 2,3-dichloro-alpha,alpha,alpha-trifluorotoluene (trace-0.005 ppm), 2,6-dichlorotoluene (not quantitated), and 2,4,5-trichlorotoluene (0.31 ppm was found in the only sample quantitated). Other isomers of tri- and tetrachloro-alpha,alpha,alpha-trifluorotoluene and di-,tri-, and tetrachlorotoluene were also present in these samples. Recoveries of the specific chlorinated trifluorotoluenes identified in these samples ranged from 86 to 108%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,4-Dichlorbenzotrifluorid, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.