Merck
  • Remodeling the integration of lipid metabolism between liver and adipose tissue by dietary methionine restriction in rats.
Remodeling the integration of lipid metabolism between liver and adipose tissue by dietary methionine restriction in rats.
Diabetes (2013-06-27)
Barbara E Hasek, Anik Boudreau, Jeho Shin, Daorong Feng, Matthew Hulver, Nancy T Van, Amanda Laque, Laura K Stewart, Kirsten P Stone, Desiree Wanders, Sujoy Ghosh, Jeffrey E Pessin, Thomas W Gettys
ZUSAMMENFASSUNG
Dietary methionine restriction (MR) produces an integrated series of biochemical and physiological responses that improve biomarkers of metabolic health, limit fat accretion, and enhance insulin sensitivity. Using transcriptional profiling to guide tissue-specific evaluations of molecular responses to MR, we report that liver and adipose tissue are the primary targets of a transcriptional program that remodeled lipid metabolism in each tissue. The MR diet produced a coordinated downregulation of lipogenic genes in the liver, resulting in a corresponding reduction in the capacity of the liver to synthesize and export lipid. In contrast, the transcriptional response in white adipose tissue (WAT) involved a depot-specific induction of lipogenic and oxidative genes and a commensurate increase in capacity to synthesize and oxidize fatty acids. These responses were accompanied by a significant change in adipocyte morphology, with the MR diet reducing cell size and increasing mitochondrial density across all depots. The coordinated transcriptional remodeling of lipid metabolism between liver and WAT by dietary MR produced an overall reduction in circulating and tissue lipids and provides a potential mechanism for the increase in metabolic flexibility and enhanced insulin sensitivity produced by the diet.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Methionin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Methionin
Sigma-Aldrich
L-Methionin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionin, reagent grade, ≥98% (HPLC)
Supelco
L-Methionin, Pharmaceutical Secondary Standard; Certified Reference Material
Methionin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Methionin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland