Merck
  • Monoamine oxidase-A knockdown in human neuroblastoma cells reveals protection against mitochondrial toxins.
Monoamine oxidase-A knockdown in human neuroblastoma cells reveals protection against mitochondrial toxins.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2013-09-21)
Julia C Fitzgerald, Aslihan Ugun-Klusek, George Allen, Luigi A De Girolamo, Iain Hargreaves, Christoph Ufer, Andrey Y Abramov, E Ellen Billett
ZUSAMMENFASSUNG
The study examined how the mitochondrial enzyme monoamine oxidase-A (MAO-A), which produces hydrogen peroxide as a catalytic by-product, influences death and survival mechanisms. Targeted microRNA (miRNA) was used to stably knock down MAO-A mRNA, protein, and catalytic activity by 60-70% in SH-SY5Y human neuroblastoma cells. The effects of MAO-A knockdown (KD) on ATP, oxidative stress, electron transport chain, and survival following exposure to mitochondrial toxins were assessed. In control cells, complex I inhibition resulted in caspase-mediated cell death linked with ROS production and reduced ATP, followed by up-regulation of MAO-A mRNA, protein, and enzyme activity levels. Inhibition of complex III and IV resulted in a similar increase in MAO-A expression, while up-regulation of MAO-A was lower following complex II inhibition. MAO-A KD decreased basal reactive oxygen species levels by 50% and increased levels of ATP and reduced glutathione and Bcl-2. MAO-A KD specifically increased the activity of complex I but had no effect on complex II-IV activities. Furthermore, MAO-A KD protected against inhibitors of complex I, III, and IV. In summary, endogenous MAO-A levels influence mitochondrial function, notably complex I activity, and MAO-A may be a target for protection against neurodegenerative conditions that involve oxidative stress and mitochondrial dysfunction as underlying pathogenic factors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoamine Oxidase B human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Monoaminoxidase A human, recombinant, expressed in baculovirus infected BTI insect cells