Merck
  • Artificial sweeteners stimulate adipogenesis and suppress lipolysis independently of sweet taste receptors.
Artificial sweeteners stimulate adipogenesis and suppress lipolysis independently of sweet taste receptors.
The Journal of biological chemistry (2013-09-27)
Becky R Simon, Sebastian D Parlee, Brian S Learman, Hiroyuki Mori, Erica L Scheller, William P Cawthorn, Xiaomin Ning, Katherine Gallagher, Björn Tyrberg, Fariba M Assadi-Porter, Charles R Evans, Ormond A MacDougald
ZUSAMMENFASSUNG
G protein-coupled receptors mediate responses to a myriad of ligands, some of which regulate adipocyte differentiation and metabolism. The sweet taste receptors T1R2 and T1R3 are G protein-coupled receptors that function as carbohydrate sensors in taste buds, gut, and pancreas. Here we report that sweet taste receptors T1R2 and T1R3 are expressed throughout adipogenesis and in adipose tissues. Treatment of mouse and human precursor cells with artificial sweeteners, saccharin and acesulfame potassium, enhanced adipogenesis. Saccharin treatment of 3T3-L1 cells and primary mesenchymal stem cells rapidly stimulated phosphorylation of Akt and downstream targets with functions in adipogenesis such as cAMP-response element-binding protein and FOXO1; however, increased expression of peroxisome proliferator-activated receptor γ and CCAAT/enhancer-binding protein α was not observed until relatively late in differentiation. Saccharin-stimulated Akt phosphorylation at Thr-308 occurred within 5 min, was phosphatidylinositol 3-kinase-dependent, and occurred in the presence of high concentrations of insulin and dexamethasone; phosphorylation of Ser-473 occurred more gradually. Surprisingly, neither saccharin-stimulated adipogenesis nor Thr-308 phosphorylation was dependent on expression of T1R2 and/or T1R3, although Ser-473 phosphorylation was impaired in T1R2/T1R3 double knock-out precursors. In mature adipocytes, artificial sweetener treatment suppressed lipolysis even in the presence of forskolin, and lipolytic responses were correlated with phosphorylation of hormone-sensitive lipase. Suppression of lipolysis by saccharin in adipocytes was also independent of T1R2 and T1R3. These results suggest that some artificial sweeteners have previously uncharacterized metabolic effects on adipocyte differentiation and metabolism and that effects of artificial sweeteners on adipose tissue biology may be largely independent of the classical sweet taste receptors, T1R2 and T1R3.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin-Esterase aus Pseudomonas fluorescens, lyophilized powder, ≥10,000 units/g protein
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Saccharin, ≥99%
Supelco
Mettler-Toledo-Kalibriersubstanz ME 51143091, Saccharin, traceable to primary standards (LGC)
Sigma-Aldrich
Triglycerid- und freie Glycerin-Kits und -Reagenzien, 1 kit sufficient for 1000 reactions
Sigma-Aldrich
Cholesterin-Esterase aus Rinderpankreas, lyophilized powder, ≥200 units/g protein
Sigma-Aldrich
Cholesterin-Esterase aus Pseudomonas sp., lyophilized powder, ≥200,000 units/g protein
Sigma-Aldrich
Cholesterinesterase aus Schweinepankreas, lyophilized, powder, white, ~35 U/mg
Sigma-Aldrich
NKH 477, ≥98% (HPLC), powder