Merck
  • Perfusion characteristics of Moyamoya disease: an anatomically and clinically oriented analysis and comparison.
Perfusion characteristics of Moyamoya disease: an anatomically and clinically oriented analysis and comparison.
Stroke (2013-11-07)
Gerrit Alexander Schubert, Marcus Czabanka, Marcel Seiz, Peter Horn, Peter Vajkoczy, Claudius Thomé
ZUSAMMENFASSUNG
Moyamoya disease (MMD) is characterized by unique angiographic features of collateralization. However, a detailed quantification as well as comparative analysis with cerebrovascular atherosclerotic disease (CAD) and healthy controls have not been performed to date. We reviewed 67 patients with MMD undergoing Xenon-enhanced computed tomography, as well as 108 patients with CAD and 5 controls. In addition to cortical, central, and infratentorial regions of interest, particular emphasis was put on regions that are typically involved in MMD (pericallosal territory, basal ganglia). Cerebral blood flow (CBF), cerebrovascular reserve capacity (CVRC), and hemodynamic stress distribution were calculated. MMD is characterized by a significant, ubiquitous decrease in CVRC and a cortical but not pericallosal decrease in CBF when compared with controls. Baseline perfusion is maintained within the basal ganglia, and hemodynamic stress distribution confirmed a relative preservation of central regions of interest in MMD, indicative for its characteristic proximal collateralization pattern. In MMD and CAD, cortical and central CBF decreased significantly with age, whereas CVRC and hemodynamic stress distribution are relatively unaffected by age. No difference in CVRC of comparable regions of interest was seen between MMD and CAD, but stress distribution was significantly higher in MMD, illustrating the functionality of the characteristic rete mirabilis. Our data provide quantitative support for a territory-specific perfusion pattern that is unique for MMD, including central preservation of CBF compared with controls and patients with CAD. This correlates well with its characteristic feature of proximal collateralization. CVRC and hemodynamic stress distribution seem to be more robust parameters than CBF alone for assessment of disease severity.