Merck
  • Comparison of microleakage of different margin types around Class V resin restorations in primary teeth.
Comparison of microleakage of different margin types around Class V resin restorations in primary teeth.
European journal of paediatric dentistry : official journal of European Academy of Paediatric Dentistry (2013-12-04)
T C Lopes Coutinho, M Almeida Tostes
ZUSAMMENFASSUNG
This in vitro study compared the effect of a concave with a straight-bevelled cavity margin on the microleakage of Class V composite resin restorations in primary teeth. Standardised Class V cavity preparations were made in vitro on the buccal (all margins placed in enamel) and on the lingual (margins placed in enamel and cementum) surfaces of 20 sound primary molars. The teeth were randomly assigned to two groups of 10 each: in Group 1, a concave bevel was made with a high-speed No. 04 tungsten carbide bur and in Group 2, a straight bevel was made with a high-speed No. 556 tungsten carbide bur. The teeth were restored incrementally with Adper Single Bond 2 (3M) adhesive and Filtek Z 350 (3M) composite resin. All specimens were subsequently thermocycled and immersed in 50% silver nitrate solution. Microleakage of the restorations was then assessed by silver penetration. A grading scale of 0 to 4 was used as the scoring criterion. At the enamel margins no statistically significant differences were observed between the two groups (p>0.05). Occlusal walls in enamel, in both groups, exhibited less leakage than the cervical walls in cementum (p<0.01) and Group 1 showed better results than Group 2 in decreasing microleakage at the cementum margins (p <0.05). Based on the results, it was concluded that concave-beveled cavity preparations may reduce but did not totally eliminate microleakage at the cementum margins of Class V composite resin restorations in primary teeth.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphorsäure, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphorsäure, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Wolfram(IV)-carbid, powder, 2 μm, ≥99%
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.