Merck
Attenuation of heavy metals by geosynthetics in the coal gangue-filled columns.
Environmental technology (2014-02-18)
Ping Wang, Zhenqi Hu, Peijun Wang
ZUSAMMENFASSUNG
In the subsided areas backfilled with coal gangue, an issue of continuing environmental concern is the migration of hazardous metals to the subsurface soil and groundwater. As an effective isolation material, geosynthetics have been scarcely applied into mining areas reclamation of China. This paper describes research aimed at characterizing the behaviours of different geosynthetics in the leaching columns filled with coal gangues. Four types of geosynthetics were selected: fibres needle-punched nonwoven geotextiles, high-density polyethylene, needle-punched Na-bentonite geosynthetic clay liner (GCL-NP) and Na-bentonite geosynthetic-overbited film. Heavy metals were significantly attenuated and by monitoring aqueous solutions in the whole percolation period, negative correlation was found between pH value and concentration of heavy metals. Generally, GCL-NP showed comparatively better effects on attenuating the migration of heavy metals. According to the meta-analysis of heavy metals present in the leachates and retained in the columns, geosynthetics have good capabilities of sorption and retardation, which can delay the breakthrough time of heavy metals and retard the accumulation in the subsurface. Future research will use X-ray diffraction and micro-imaging (electron microprobe and scanning electron microscopy) to further explain retention mechanisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bentonit
Sigma-Aldrich
Polyethylen, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylen, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Nanoton, hydrophiles Bentonit
Sigma-Aldrich
Polyethylen, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylen, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylen, High density, melt index 12 g/10 min (190 °C/2.16kg)
Polyethylen (LDPE), ERM®, certified reference material
Sigma-Aldrich
Polyethylen, Medium density
Sigma-Aldrich
Polyethylen, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylen, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylen, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Supelco
Polyethylen, analytical standard, suitable for gel permeation chromatography (GPC), 2,000