Merck
Genome-wide miRNA-profiling of aflatoxin B1-induced hepatic injury using deep sequencing.
Toxicology letters (2014-01-30)
Weiqiang Yang, Junwei Lian, Youjun Feng, Swaminath Srinivas, Zhenni Guo, Hong Zhong, Zhenhong Zhuang, Shihua Wang
ZUSAMMENFASSUNG
Aflatoxin B1 is a potent carcinogen which can induce** hepatocellular carcinoma (HCC) in mammals. Though microRNAs are known to play important roles in tumorigenesis, the functional complexity of microRNAs in AFB1-induced hepatocellular tumorigenesis has not yet been elucidated. Here, we applied Illumina deep sequencing technology for high-throughput profiling of microRNA in rat liver tissue before and after treatment with aflatoxin B1. Analysis of mature miRNAs from different arms of pre-miRNAs allowed us to identify the predominant form of miRNA. We studied the differential expression profile of miRNAs in two libraries, identifying several cancer-related microRNAs which exhibit abnormal expression. KEGG analysis indicated that predicted target genes of differentially expressed miRNAs are involved in cancer-related pathways. Bioinformatic analysis predicted 16 potential novel miRNAs. Our work provides new insights at the miRNA level into AFB1-induced hepatic injury which may lead to HCC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aflatoxin B1, from Aspergillus flavus
Supelco
Aflatoxin B1 -Lösung, 2 μg/mL in acetonitrile, analytical standard
Aflatoxin B1 -Lösung, 3.79 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin B1 -Lösung, 3 μg/mL in benzene:acetonitrile (98:2), analytical standard