Merck
  • The kynurenine pathway and quinolinic acid: pivotal roles in HIV associated neurocognitive disorders.
The kynurenine pathway and quinolinic acid: pivotal roles in HIV associated neurocognitive disorders.
The FEBS journal (2012-01-21)
Apsara Kandanearatchi, Bruce J Brew
ZUSAMMENFASSUNG
This brief review will first consider HIV associated neurocognitive disorder followed by the current understanding of its neuropathogenesis. Against this background the role of the kynurenine pathway will be detailed. Evidence both direct and indirect will be discussed for involvement of the kynurenine pathway at each step in the neuropathogenesis of HIV associated neurocognitive disorder.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,3-Pyridindicarboxylsäure, 99%