Merck
Nefiracetam, a novel cognition-enhancing agent. An introductory overview.
Arzneimittel-Forschung (1994-02-01)
M Tanaka, K Takasuna, S Takayama
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nefiracetam, solid