Merck
  • Effects of elevated temperature on the toxicity of copper and oxytetracycline in the marine model, Euplotes crassus: a climate change perspective.
Effects of elevated temperature on the toxicity of copper and oxytetracycline in the marine model, Euplotes crassus: a climate change perspective.
Environmental pollution (Barking, Essex : 1987) (2014-08-29)
A Gomiero, A Viarengo
ZUSAMMENFASSUNG
Trace metals and broad-spectrum antibiotic drugs are common environmental contaminants, the importance of which is increasing due to global climate change-related effects. In the present study, the biological model organism E. crassus was first acclimated to five temperatures, from 25 °C to 33 °C, followed by exposure to nominal concentrations of copper, the antibiotic model compound oxytetracycline and mixtures of both, at increasing thermal conditions. Variations of temperature-related toxicity were assessed by two high-level endpoint tests, survival and replication rates, and two sublethal parameters: endocytosis rate and lysosomal membrane stability. The selected toxicants presented opposite behaviours as the protozoa's survival rates increased following an increasing thermal gradient in the oxytetracycline-related treatments, and a decline of tolerance in metal-related treatments was observed. Results of tests combining binary mixtures of tested toxicants showed a complex pattern of responses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kupfer, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Kupfer, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Oxytetracyclin -hydrochlorid
Sigma-Aldrich
Kupfer, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Kupfer, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Kupfer, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Kupfer, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Kupfer, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Kupfer, nanopowder, 60-80 nm particle size (APS), ≥99.5% trace metals basis
Sigma-Aldrich
Kupfer, ACS reagent, granular, 10-40 mesh, ≥99.90%
Supelco
Oxytetracyclin-Hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Oxytetracyclin Dihydrat, Assay (anhydrous basis) 95.0%-102.0%
Sigma-Aldrich
Kupfer, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Kupfer, beads, 2-8 mm, 99.9995% trace metals basis
Supelco
Oxytetracyclin -hydrochlorid, VETRANAL®, analytical standard
Sigma-Aldrich
Kupfer, foil, thickness 1.0 mm, 99.999% trace metals basis
Kupfer, mesh, 100x100mm, nominal aperture 0.14mm, thickness 0.25mm, wire diameter 0.115mm, 100x100 wires/inch, open area 30.3%, plain weave mesh
Sigma-Aldrich
Kupfer, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Kupfer, wire, diam. 0.25 mm, 99.999% trace metals basis
USP
Oxytetracyclin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Kupfer, shot, −3-+14 mesh, 99%
Millipore
Oxytetra-Selektivzusatz, suitable for microbiology
Sigma-Aldrich
Kupfer, beads, 2-8 mm, ≥99.99% trace metals basis
Kupfer, wire reel, 100m, diameter 0.05mm, annealed, 99.9%
USP
Oxytetracyclin -hydrochlorid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Kupfer, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Kupfer, wire, diam. 0.64 mm, 99.995% trace metals basis
Oxytetracyclin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Kupfer, sheet, 100x100mm, thickness 6.35mm, 99.95+%
Kupfer, rod, 500mm, diameter 2.0mm, as drawn, 99.99+%