Merck
Inter-individual variation in midazolam clearance in children.
Archives of disease in childhood (2014-10-05)
Mohammed I Altamimi, Helen Sammons, Imti Choonara
ZUSAMMENFASSUNG
To determine the extent of inter-individual variation in clearance of midazolam in children and establish which factors are responsible for this variation. A systematic literature review was performed to identify papers describing the clearance of midazolam in children. The following databases were searched: Medline, Embase, International Pharmaceutical Abstracts, CINAHL and Cochrane Library. From the papers, the range in plasma clearance and the coefficient of variation (CV) in plasma clearance were determined. 25 articles were identified. Only 13 studies gave the full range of clearance values for individual patients. The CV was greater in critically ill patients (18%-170%) than non-critically ill patients (13%-54%). Inter-individual variation was a major problem in all age groups of critically ill patients. The CV was 72%-106% in preterm neonates, 18%-73% in term neonates, 31%-130% in infants, 21%-170% in children and 47%-150% in adolescents. The mean clearance was higher in children (1.1-16.7 mL/min/kg) than in neonates (0.78-2.5 mL/min/kg). Large inter-individual variation was seen in midazolam clearance values in critically ill neonates, infants, children and adolescents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Midazolam -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Midazolam for system suitability, European Pharmacopoeia (EP) Reference Standard