Merck
  • The neurofibromatosis 2 tumor suppressor protein interacts with hepatocyte growth factor-regulated tyrosine kinase substrate.
The neurofibromatosis 2 tumor suppressor protein interacts with hepatocyte growth factor-regulated tyrosine kinase substrate.
Human molecular genetics (2000-06-22)
D R Scoles, D P Huynh, M S Chen, S P Burke, D H Gutmann, S M Pulst
ZUSAMMENFASSUNG
The neurofibromatosis 2 tumor suppressor protein schwannomin/merlin is commonly mutated in schwannomas and meningiomas. Schwannomin, a member of the 4.1 family of proteins, which are known to link the cytoskeleton to the plasma membrane, has little known function other than its ability to suppress tumor growth. Using yeast two-hybrid interaction cloning, we identified the HGF-regulated tyrosine kinase substrate (HRS) as a schwannomin interactor. We verified the interaction by both immunoprecipitation of endogenous HRS with endogenous schwannomin in vivo as well as by using bacterially purified HRS and schwannomin in vitro. We narrowed the regions of interaction to include schwannomin residues 256-579 and HRS residues from 480 to the end of either of two HRS isoforms. Schwannomin molecules with a L46R, L360P, L535P or Q538P missense mutation demonstrated reduced affinity for HRS binding. As HRS is associated with early endosomes and may mediate receptor translocation to the lysosome, we demonstrated that schwannomin and HRS co-localize at endosomes using the early endosome antigen 1 in STS26T Schwann cells by indirect immunofluorescence. The identification of schwannomin as a HRS interactor implicates schwannomin in HRS-mediated cell signaling.