Merck
  • LC-MS/MS quantitative determination of Tetrapterys mucronata alkaloids, a plant occasionally used in ayahuasca preparation.
LC-MS/MS quantitative determination of Tetrapterys mucronata alkaloids, a plant occasionally used in ayahuasca preparation.
Phytochemical analysis : PCA (2015-01-27)
M M F Queiroz, G Marti, E F Queiroz, L Marcourt, I Castro-Gamboa, V S Bolzani, J-L Wolfender
ZUSAMMENFASSUNG
Tetrapterys mucronata Cav. (Malpighiaceae) is a plant used in some regions of Brazil in the preparation of ayahuasca. To determine the content of the main tryptamine alkaloids in the stem bark of T. mucronata Cav. and assess their possible toxic and hallucinogenic properties based on the doses found in a water decoction that mimics the ayahuasca preparation. Four alkaloids previously described for their toxic and hallucinogenic properties were quantitated by multiple reaction monitoring HPLC combined with electrospray ionisation and tandem MS (HPLC-ESI/MS/MS) in the water decoction and ethanolic extracts from the bark of T. mucronata. Exhaustive extraction of the stem barks with ethanol revealed the following alkaloid levels: bufotenine (1) 3.26 ± 0.31 mg/g, 5-methoxy-N-methyltryptamine (2) 0.88 ± 0.08 mg/g, 5-methoxy-bufotenine (3) 3.07 ± 0.22 mg/g and 2-methyl-6-methoxy-1,2,3,4-tetrahydro-β-carboline (4) 0.14 ± 0.004 mg/g. The water decoction presented slightly lower levels, ranging between 2.32 ± 0.14, 0.50 ± 0.04, 1.53 ± 0.09 and 0.10 ± 0.01 mg/g for (1), (2), (3) and (4) respectively. The HPLC-ESI/MS/MS quantitation revealed significant alkaloid levels, in particular for bufotenine and 5-methoxy-bufotenine. As such compounds are known for their toxic and hallucinogenic properties, these results indicate that the consumption of this plant as an ingredient in ayahuasca preparations may present a risk to consumers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ameisensäure, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, ≥99.9% (GC)
Sigma-Aldrich
Ameisensäure, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Natriumformiat, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitril, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Natriumformiat, reagent grade, 97%
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumformiat, BioUltra, ≥99.0% (NT)
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Natriumformiat, 99.998% trace metals basis
Sigma-Aldrich
Acetonitril, ≥99.5% (GC)
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O
Leucin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Leucin, ≥99% (HPLC)
Supelco
Natriumformiat, analytical standard
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material