Merck
  • Nebivolol potentiates the efficacy of PDE5 inhibitors to relax corpus cavernosum and penile arteries from diabetic patients by enhancing the NO/cGMP pathway.
Nebivolol potentiates the efficacy of PDE5 inhibitors to relax corpus cavernosum and penile arteries from diabetic patients by enhancing the NO/cGMP pathway.
The journal of sexual medicine (2014-05-31)
Juan I Martínez-Salamanca, José M La Fuente, José Cardoso, Argentina Fernández, Pedro Cuevas, Harold M Wright, Javier Angulo
ZUSAMMENFASSUNG
The efficacy of oral pharmacotherapy for erectile dysfunction (ED) (i.e., type 5 phosphodiesterase[PDE5] inhibitors) is significantly reduced in diabetic patients. Nebivolol is a selective β1-blocker used for treatinghy pertension that has been shown to increase the efficacy of sildenafil to reverse ED in diabetic rats. To evaluate the effects of nebivolol on the efficacy of the PDE5 inhibitors, sildenafil, tadalafil, and vardenafil to relax human corpus cavernosum (HCC) and vasodilate human penile resistance arteries (HPRA) from diabetic patients with ED (DMED). The influence of nebivolol on the capacity of these three PDE5 inhibitors to stimulate cyclic guanosine monophosphate (cGMP) production in HCC was also evaluated. HCC and HPRA were obtained from organ donors without ED (NEND; n = 18) or patients with diabetes undergoing penile prosthesis implantation (DMED; n = 19). Relaxations of HCC strips and HPRA to sildenafil,tadalafil, and vardenafil were evaluated in organ chambers and wire myographs. cGMP content in HCC was determined by ether extraction and quantification by ELISA. Effects of nebivolol on PDE5 inhibitor-induced relaxation of HCC, vasodilation ofHPRA and cGMP accumulation in HCC. Treatment with nebivolol (1 μM) significantly potentiated sildenafil-, tadalafil- and vardenafil-induced relaxations of HCC and vasodilations of HPRA from both NEND and DMED. Enhancement of relaxant capacity by nebivolol resulted in reversion of the impairment of PDE5 inhibition-induced responses in DMED and it was accompanied by enhancing the ability of PDE5 inhibitors to increase cGMP in HCC restoring reduced cGMP levelsin HCC from DMED. Nebivolol potentiated the capacity of PDE5 inhibitors to relax vascular structures of erectile tissue from diabetic patients by enhancing the nitric oxide (NO)/cGMP pathway in these tissues. These effects suggest a potential therapeutic utility of nebivolol as an adjunct to PDE5 inhibitors for the treatment of ED associated with diabetes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Acetylcholinchlorid, ≥99% (TLC)
Supelco
D-Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
USP
Natriumbicarbonat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Acetylcholinchlorid, suitable for cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Guanosin-3′,5′-cyclophosphat Natriumsalz, ≥99% (HPLC), powder
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.