Merck
  • Slow-releasing rapamycin-coated bionic peripheral nerve scaffold promotes the regeneration of rat sciatic nerve after injury.
Slow-releasing rapamycin-coated bionic peripheral nerve scaffold promotes the regeneration of rat sciatic nerve after injury.
Life sciences (2014-12-23)
Tan Ding, Chao Zhu, Jun-Bin Yin, Ting Zhang, Ya-Cheng Lu, Jun Ren, Yun-Qing Li
ZUSAMMENFASSUNG
To investigate the effect of locally slow-released rapamycin (RAPA) from the bionic peripheral nerve scaffold on rat sciatic nerve regeneration in the early phase of nerve injury. Slow-releasing RAPA-polyhydroxy alcohol (PLGA) microspheres were prepared and tested for microsphere diameter and slow-release effect in vitro after loading onto nerve scaffold. A total of 48 male SD rats were randomly divided into control group and 3 experimental groups as follows: group 1: RAPA-PLGA scaffold; group 2: RAPA scaffold; and group 3: scaffold alone. In the control group, a 15mm sciatic nerve was excised and religated reversely. In the experimental groups, the scaffolds were used to bridge a defect of 15mm sciatic nerve. The outcome of nerve regeneration was evaluated using neurophysiological and neuromuscular morphological techniques. The RAPA-PLGA microspheres displayed a smooth exterior. The slow-release of RAPA in group 1 lasted for 14days. The sciatic nerve function index (SFI) and electrophysiological and morphological features were examined 12weeks after the surgery in all groups to reveal various degrees of ipsilateral sciatic nerve regeneration. The SFI values at 12weeks showed no significant difference between the RAPA-PLGA scaffold and control groups; morphological observations revealed that the outcomes of nerve regeneration in the above 2 groups were similar and significantly better than those in the RAPA scaffold and scaffold alone groups. RAPA-PLGA microsphere-loaded bionic peripheral nerve scaffold gradually released RAPA locally in the early phase of sciatic nerve regeneration, reduced the secondary nerve injury, and evidently promoted the regeneration of peripheral nerve.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, ≥99.9% (GC)
Sigma-Aldrich
Dichlormethan, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Kollagen aus Achillessehne vom Rind, powder, suitable for substrate for collagenase
Sigma-Aldrich
Gelatine aus Rinderhaut, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Dichlormethan, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Supelco
Dichlormethan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
Dichlormethan, analytical standard
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, ≥99.5% (GC)
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Dichlormethan, Selectophore, ≥99.5%