Merck
In vitro and in vivo effects of tamoxifen against larval stage Echinococcus granulosus.
Antimicrobial agents and chemotherapy (2014-06-18)
María Celeste Nicolao, María Celina Elissondo, Guillermo M Denegri, Alejandra B Goya, Andrea C Cumino
ZUSAMMENFASSUNG
Cystic echinococcosis is a zoonotic infection caused by the larval stage of the cestode Echinococcus granulosus. Chemotherapy currently employs benzimidazoles; however, 40% of cases do not respond favorably. With regard to these difficulties, novel therapeutic tools are needed to optimize treatment in humans. The aim of this work was to explore the in vitro and in vivo effects of tamoxifen (TAM) against E. granulosus. In addition, possible mechanisms for the susceptibility of TAM are discussed in relation to calcium homeostasis, P-glycoprotein inhibition, and antagonist effects on a putative steroid receptor. After 24 h of treatment, TAM, at a low micromolar concentration range (10 to 50 μM), inhibited the survival of E. granulosus protoscoleces and metacestodes. Moreover, we demonstrated the chemotherapeutic and chemopreventive pharmacological effects of the drug. At a dose rate of 20 mg/kg of body weight, TAM induced protection against the infection in mice. In the clinical efficacy studies, a reduction in cyst weight was observed after the administration of 20 mg/kg in mice with cysts developed during 3 or 6 months, compared to that of those collected from control mice. Since the collateral effects of high TAM doses have been largely documented in clinical trials, the use of low doses of this drug as a short-term therapy may be a novel alternative approach for human cystic echinococcosis treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Tamoxifen -citrat (Salz), ≥99%
Sigma-Aldrich
Calcein-AM -Lösung, 4 mM in DMSO, ≥90% (HPLC), solution
Tamoxifencitrat für Performance-Test, European Pharmacopoeia (EP) Reference Standard
Tamoxifencitrat, European Pharmacopoeia (EP) Reference Standard