Merck
  • Functional analysis of iPSC-derived myocytes from a patient with carnitine palmitoyltransferase II deficiency.
Functional analysis of iPSC-derived myocytes from a patient with carnitine palmitoyltransferase II deficiency.
Biochemical and biophysical research communications (2014-05-02)
Tetsuhiko Yasuno, Kenji Osafune, Hidetoshi Sakurai, Isao Asaka, Akihito Tanaka, Seiji Yamaguchi, Kenji Yamada, Hirofumi Hitomi, Sayaka Arai, Yuko Kurose, Yasuki Higaki, Mizuki Sudo, Soichi Ando, Hitoshi Nakashima, Takao Saito, Hidetoshi Kaneoka
ZUSAMMENFASSUNG
Carnitine palmitoyltransferase II (CPT II) deficiency is an inherited disorder involving β-oxidation of long-chain fatty acids (FAO), which leads to rhabdomyolysis and subsequent acute renal failure. The detailed mechanisms of disease pathogenesis remain unknown; however, the availability of relevant human cell types for investigation, such as skeletal muscle cells, is limited, and the development of novel disease models is required. We generated human induced pluripotent stem cells (hiPSCs) from skin fibroblasts of a Japanese patient with CPT II deficiency. Mature myocytes were differentiated from the patient-derived hiPSCs by introducing myogenic differentiation 1 (MYOD1), the master transcriptional regulator of myocyte differentiation. Using an in vitro acylcarnitine profiling assay, we investigated the effects of a hypolipidemic drug, bezafibrate, and heat stress on mitochondrial FAO in CPT II-deficient myocytes and controls. CPT II-deficient myocytes accumulated more palmitoylcarnitine (C16) than did control myocytes. Heat stress, induced by incubation at 38°C, leads to a robust increase of C16 in CPT II-deficient myocytes, but not in controls. Bezafibrate reduced the amount of C16 in control and CPT II-deficient myocytes. In this study, we induced differentiation of CPT II-deficient hiPSCs into mature myocytes in a highly efficient and reproducible manner and recapitulated some aspects of the disease phenotypes of CPT II deficiency in the myocyte disease models. This approach addresses the challenges of modeling the abnormality of FAO in CPT II deficiency using iPSC technology and has the potential to revolutionize translational research in this field.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Palmitinsäure, ≥99%
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Palmitinsäure, BioXtra, ≥99%
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Supelco
Palmitinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
L-Carnitin Inneres Salz, synthetic, ≥98%
Sigma-Aldrich
Palmitinsäure, ≥98%, FCC, FG
USP
Palmitinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Palmitinsäure, analytical standard
Sigma-Aldrich
Palmitinsäure, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Bezafibrat, ≥98%, solid
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
USP
Levocarnitin, United States Pharmacopeia (USP) Reference Standard
Palmitinsäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Palmitinsäure, natural, 98%, FG
Supelco
Calcium Ionenlösung für ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Supelco
Palmitinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
FGF-2 human, recombinant, expressed in insect cells, ≥85% (SDS-PAGE)
Levocarnitin, European Pharmacopoeia (EP) Reference Standard
Bezafibrat, European Pharmacopoeia (EP) Reference Standard