Merck
Screening of DUB activity and specificity by MALDI-TOF mass spectrometry.
Nature communications (2014-08-28)
Maria Stella Ritorto, Richard Ewan, Ana B Perez-Oliva, Axel Knebel, Sara J Buhrlage, Melanie Wightman, Sharon M Kelly, Nicola T Wood, Satpal Virdee, Nathanael S Gray, Nicholas A Morrice, Dario R Alessi, Matthias Trost
ZUSAMMENFASSUNG
Deubiquitylases (DUBs) are key regulators of the ubiquitin system which cleave ubiquitin moieties from proteins and polyubiquitin chains. Several DUBs have been implicated in various diseases and are attractive drug targets. We have developed a sensitive and fast assay to quantify in vitro DUB enzyme activity using matrix-assisted laser desorption/ionization time-of-flight (MALDI-TOF) mass spectrometry. Unlike other current assays, this method uses unmodified substrates, such as diubiquitin topoisomers. By analysing 42 human DUBs against all diubiquitin topoisomers we provide an extensive characterization of DUB activity and specificity. Our results confirm the high specificity of many members of the OTU and JAB/MPN/Mov34 metalloenzyme DUB families and highlight that all USPs tested display low linkage selectivity. We also demonstrate that this assay can be deployed to assess the potency and specificity of DUB inhibitors by profiling 11 compounds against a panel of 32 DUBs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, ≥99.9% (GC)
Sigma-Aldrich
Acetonitril, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
α-Cyano-4-hydroxyzimtsäure, suitable for MALDI-TOF MS
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
α-Cyano-4-hydroxyzimtsäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
2,4,6-Trimethylpyridin, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
α-Cyano-4-hydroxyzimtsäure, 99%
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
α-Cyano-4-hydroxyzimtsäure, ≥98% (TLC), powder
Sigma-Aldrich
Pimozid
Sigma-Aldrich
Acetonitril, ≥99.5% (GC)
Sigma-Aldrich
PR-619, ≥95% (HPLC)
Sigma-Aldrich
α-Cyano-4-hydroxyzimtsäure, suitable for matrix substance for MALDI-MS, Ultra pure
Sigma-Aldrich
2,4,6-Trimethylpyridin, puriss. p.a., 99% (GC)
Sigma-Aldrich
α-Cyano-4-hydroxyzimtsäure, 97%
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, ≥99.8% (GC)
Pimozid, European Pharmacopoeia (EP) Reference Standard