Merck
CD226 as a genetic adjuvant to enhance immune efficacy induced by Ag85A DNA vaccination.
International immunopharmacology (2015-01-15)
Yan Li, Fangli Yang, Junfeng Zhu, Lixuan Sang, Xue Han, Danan Wang, Fengping Shan, Shengjun Li, Xun Sun, Changlong Lu
ZUSAMMENFASSUNG
Antigen-85A (Ag85A) is one of the major proteins secreted by Mycobacterium tuberculosis. Many studies on animal models have shown that vaccination with the recombinant Ag85A-DNA or Ag85A protein induces powerful immune response. However, these vaccines cannot generate sufficient protective immunity in the systemic compartment. CD226, a member of the immunoglobulin superfamily, is expressed in the majority of NK cells, T cells, monocytes, and platelets, and can be served as a co-stimulator that contributes to multiple innate and adaptive responses. However, there has been no study where either CD226 protein or DNA has been used as an adjuvant for vaccine development. The aim of this study was to develop a novel Ag85A DNA vaccine with CD226 as the genetic adjuvant to increase the immune efficacy induced by Ag85A. Oral vaccination with pcDNA3.1-Ag85A-CD226 DNA induced potent immune responses in mice. CD226 was an effective genetic adjuvant that improved the immune efficacy induced by Ag85A and enhanced the activity of cytotoxic T lymphocytes (CTL) and NK cells in mice. Th1 dominant cytokines (i.e. IL-2, IFN-γ and TNF-α), cellular immunity (i.e. CD4(+)IFN-γ(+)T cells and CD8(+)IFN-γ(+)T cells in splenocytes) and MLNs were also significantly elevated by pcDNA3.1-Ag85A-CD226 DNA vaccination. Our results suggest that CD226 is an effective adjuvant to enhance the immune efficacy induced by Ag85A. Our findings provide a new strategy for the development of a DNA vaccine co-expressing Ag85A and CD226.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Natrium DL-Laktat -Lösung, syrup, BioReagent, 60 % (w/w), synthetic, suitable for cell culture
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥99%
Sigma-Aldrich
Aphidicolin, ≥98% (HPLC), powder
Sigma-Aldrich
Natrium DL-Laktat -Lösung, syrup, 60 % (w/w), synthetic
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Natrium DL-Laktat -Lösung, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, Grade AA-1
Sigma-Aldrich
Natrium-DL-lactat, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Supelco
Aphidicolin, analytical standard