Merck
Surface selective one-step fabrication of carbon nanotube thin films with high density.
ACS nano (2014-04-02)
Jose M Lobez, Shu-Jen Han, Ali Afzali, James B Hannon
ZUSAMMENFASSUNG
Thin films of carbon nanotubes (CNTs) are fabricated from solution using a one-step directed assembly strategy. Very high surface selectivity and exceptionally high CNT densities can be observed in small features with complex shapes. This directed assembly technique makes use of minimum amounts of CNTs and low toxicity solvents, and can be applied to metallic, semiconducting and mixed CNTs for fabrication of thin films over macroscopic areas. The thin films obtained with this approach are used for thin-film transistor (TFT) fabrication, and their electrical characterization is described.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles