Merck
  • Activation of p53 by sodium selenite switched human leukemia NB4 cells from autophagy to apoptosis.
Activation of p53 by sodium selenite switched human leukemia NB4 cells from autophagy to apoptosis.
Oncology research (2013-01-01)
Zhushi Li, Kejian Shi, Liying Guan, Qian Jiang, Yang Yang, Caimin Xu
ZUSAMMENFASSUNG
It was revealed by our previous research that sodium selenite repressed autophagy accompanied by the induction of apoptosis in human leukemia NB4 cells. The inhibition of autophagy exerted a facilitative effect on apoptosis. In the present study, we further explored the mechanisms underlying the switch from autophagy to apoptosis and elucidated p53 played a key role. Selenite induced phosphorylation of p53 at the vital site Ser15 via p38MAPK and ERK. Subsequently p53 dissociated with its inhibitory protein mouse double minute 2 (MDM2). Meanwhile, the nucleolar protein B23 transferred from the nucleolus to the nucleoplasm and associated with MDM2, probably stabilizing p53. The active p53 participated in the decrease of autophagic protein Beclin-1 and LC-3, as well as activation of apoptosis-related caspases. Furthermore, in p53 mutant U937 leukemia cells, selenite could not elicit such a switch from autophagy to apoptosis, laying emphasis on the crucial role p53 played in this process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Natriumselenit, 99%
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Natriumselenit, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Natriumselenit, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
USP
Natriumbicarbonat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Acridinorange -hydrochlorid -Lösung, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumselenit, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Natriumbicarbonat, −40-+140 mesh, ≥95%
Sigma-Aldrich
Natriumbicarbonat, tested according to Ph. Eur.
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C