Merck
  • Association of GLP-1 secretion with anti-hyperlipidemic effect of ginsenosides in high-fat diet fed rats.
Association of GLP-1 secretion with anti-hyperlipidemic effect of ginsenosides in high-fat diet fed rats.
Metabolism: clinical and experimental (2014-07-26)
Can Liu, Meng-yue Hu, Mian Zhang, Feng Li, Jia Li, Ji Zhang, Ying Li, Hai-fang Guo, Ping Xu, Li Liu, Xiao-dong Liu
ZUSAMMENFASSUNG
Ginsenosides, major bioactive constituents in Panax ginseng, have been shown to exert anti-hyperlipidemia effects. However, the underlying mechanism was not well-elucidated due to the low bioavailability of ginsenosides. Glucagon-like peptide-1 (GLP-1) was considered to be a critical regulator of energy homeostasis. Our previous studies have showed that ginseng total saponins (GTS) exhibited antidiabetic effects partly via modulating GLP-1 release. The aim of this study was to investigate the potential role of GLP-1 in anti-hyperlipidemia effect of GTS in rats fed with high-fat diet. Male Sprague-Dawley rats were fed with normal diet (CON) or high-fat diet (HFD) for 4 weeks. Then, the HFD rats orally received vehicle (HFD), 150 mg/kg/day (HFD-GL) and 300 mg/kg/day of GTS (HFD-GH) for another 4 weeks, respectively. Four-week GTS treatment significantly ameliorated hyperlipidemia, decreased body fat, liver weight and improved insulin resistance. It was found that high-dose GTS treatment increased portal GLP-1 level induced by glucose loading, accompanied by increased intestinal GLP-1 content, L-cell number and prohormone convertase 3 mRNA expression. Data from NCI-H716 cells showed that both GTS and ginsenoside Rb1 significantly increased GLP-1 secretion as well as proglucagon mRNA level in NCI-H716 cells supplemented with 10% HFD-rat serum. Hyperlipidemia and insulin resistance were attenuated effectively in response to GTS treatment. These improvements may be associated with the increased secretion of GLP-1.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Iod, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Iod, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Supelco
Iod, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Iod, ≥99.99% trace metals basis
Sigma-Aldrich
Iod, 99.999% trace metals basis
Sigma-Aldrich
Iod, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Iod, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iod, puriss., ≥99.5% (RT), particles (round)
Sigma-Aldrich
Iod, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Human GCG / Glucagon ELISA Kit
Sigma-Aldrich
Mouse Granzyme B ELISA Kit, for serum, plasma and cell culture supernatant