Merck
Chemerin and prediction of Diabetes mellitus type 2.
Clinical endocrinology (2015-02-03)
Thomas Bobbert, Franziska Schwarz, Antje Fischer-Rosinsky, Lukas Maurer, Matthias Möhlig, A F H Pfeiffer, Knut Mai, Joachim Spranger
ZUSAMMENFASSUNG
Insulin resistance and subclinical inflammation are characteristics in the development of type 2 diabetes mellitus (T2DM). The adipokine chemerin has been associated with both factors. The aim of this study was to analyse whether chemerin predicts T2DM. Blood samples of 440 participants of the Metabolic-Syndrome Berlin-Potsdam (MesyBepo) follow-up study without diabetes at baseline were available for chemerin measurement. Mean follow-up of participants was 5·3 years. Glucose metabolism was analysed using oral glucose tolerance test including insulin measurements. Chemerin was measured using a commercially available ELISA. Thirty-five individuals developed T2DM during follow-up. Chemerin predicted incident T2DM (Chemerin 1. Tertile: reference, 2. Tertile: OR 2·33 [0·68-7·95]; Chemerin 3. Tertile: OR 3·42 [1·01-11·58] after adjustment for age, sex, BMI, follow-up time, HbA1c, HOMA-IR and WHR). In a secondary analysis, chemerin also predicted worsening of fasting glucose and HbA1c (adjusted for age, sex, BMI, time of follow-up, WHR, HDL cholesterol and triglycerides). Our data suggest that chemerin is a weak predictor of T2DM.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder