Merck
  • PAH/DAS covalently cross-linked layer-by-layer multilayers: a "nano-net" superstratum immobilizes nanoparticles and remains permeable to small molecules.
PAH/DAS covalently cross-linked layer-by-layer multilayers: a "nano-net" superstratum immobilizes nanoparticles and remains permeable to small molecules.
Soft matter (2015-08-04)
Qi An, Kun Nie, Yihe Zhang, Yue Wang, Yingmo Hu, Victoria Dutschk, Xinglong Luan
ZUSAMMENFASSUNG
A "nano-net" superstratum strategy is developed to stabilize layer-by-layer (LbL) films that incorporate nanoparticles. The superstratum immobilizes silica, gold, or magnetic nanoparticles and at the same time is permeable to small molecules. Unlike most strategies to stabilize LbL multilayers reported in the literature, our strategy does not directly cross-link the nanoparticles and polymers in the adjacent layer, thus circumventing the tedious processes of (surface) modification of the nanoparticles or polymers. The unique advantage of our strategy is further employed in the preparation of a model functional device, where mesoporous silica nanoparticles are held in the composite multilayers with enhanced stabilities. A model drug, methylene blue, is then loaded in large amounts due to the porous structure of the silica particles, and could be released in a delayed manner up to 55 h.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Tetraethylorthosilikat, reagent grade, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥98%
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, Molecular Biology, ≥99%
Sigma-Aldrich
Tetraethylorthosilikat, ≥99.0% (GC)
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioXtra, ≥99%
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Tetraethylorthosilikat, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetraethylorthosilicat, packaged for use in deposition systems
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%
Sigma-Aldrich
Natriumhydroxid-16O -Lösung, 20 wt. % in H216O, 99.9 atom % 16O