Merck
  • Effect of alginate coatings with cinnamon bark oil and soybean oil on quality and microbiological safety of cantaloupe.
Effect of alginate coatings with cinnamon bark oil and soybean oil on quality and microbiological safety of cantaloupe.
International journal of food microbiology (2015-09-01)
Yue Zhang, Qiumin Ma, Faith Critzer, P Michael Davidson, Qixin Zhong
ZUSAMMENFASSUNG
The quality and microbiological safety of cantaloupes can potentially be improved using antimicrobial coatings that are able to maintain effectiveness throughout storage. The objective of this work was to study the effect of coating mixtures containing sodium alginate and cinnamon bark oil (CBO) on the quality of cantaloupes and the survival of inoculated bacterial pathogens and naturally occurring yeasts and molds during ambient storage at 21 °C. Cantaloupes were dipped in mixtures containing 1% sodium alginate with or without 2% CBO and 0 or 0.5% soybean oil (SBO). Weight loss and total soluble solids content of the flesh were not significantly different among coating treatments. However, changes in color and firmness of cantaloupes were delayed to different extents after coating, most significantly for the CBO+SBO treatment. Cocktails of Salmonella enterica, Escherichia coli O157:H7, or Listeria monocytogenes inoculated on cantaloupes were reduced to the detection limit (1.3 log CFU/cm(2)) and completely inhibited during the 15-day storage by the CBO+SBO treatment, while L. monocytogenes and S. enterica reached populations of 2.9 log CFU/cm(2) and 2.4 log CFU/cm(2), respectively, on cantaloupes coated with CBO alone. Antimicrobial coatings, especially with SBO, also reduced yeast and mold counts on cantaloupes by 2.6 log CFU/cm(2). SBO improved the retention of CBO during storage suggesting it is related to the enhancement of quality and microbiological safety. Findings demonstrated the potential of the antimicrobial coating system studied to improve microbiological safety and quality of cantaloupes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Glycerin, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerin -Lösung, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Linolsäure, technical, 58-74% (GC)
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Glycerin, Vetec, reagent grade, 99%