Merck
Novel doped calcium phosphate-PMMA bone cement composites as levofloxacin delivery systems.
International journal of pharmaceutics (2015-05-24)
Ana C Matos, Catarina F Marques, Rosana V Pinto, Isabel A C Ribeiro, Lídia M Gonçalves, Mário A Vaz, J M F Ferreira, António J Almeida, Ana F Bettencourt
ZUSAMMENFASSUNG
Antibiotic-loaded acrylic bone cements (ALABCs) are well-established and cost-effective materials to control the occurrence of bone and joint infections. However, the inexistence of alternative antibiotics other than those already commercially available and the poor ability to bind to bone tissue hampering its biological function are still major drawbacks of ALABCs clinical application. The concept of this research work is to develop a novel bone cement (BC) drug delivery system composed by Mg- and Sr-doped calcium phosphate (CaP) particles as drug carriers loaded into a lactose-modified acrylic BC, which, to the best of our knowledge, has never been reported. CaP particles are known to promote bone ingrowth and current research is focused on using these carriers as antibiotic delivery systems for the treatment of bone infections, like osteomyelitis. Levofloxacin is a fluoroquinolone with anti-staphylococcal activity and adequate penetration into osteoarticular tissues and increasingly being recommended to manage bone-related infections. Also, the lactose-modified BC matrix, with a more porous structure, has already proved to enhance antibiotic release from the BC inner matrix. This novel BC composite biomaterial has shown improved mechanical integrity, biocompatibility maintenance, and sustained release of levofloxacin, with concentrations over the minimum inhibitory concentration values after a 48h while maintaining antibacterial activity over an 8-week period against Staphyloccocus aureus and Staphyloccocus epidermidis, common pathogens associated with bone infections.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
SAFC
Natriumchlorid -Lösung, 5 M
SAFC
L-Glutamin
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Strontium, granular, 99% trace metals basis
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Strontium, random pieces, 99%
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.9% trace metals basis