Merck
  • Aging rather than sun exposure is a major determining factor for the density of miR-125b-positive epidermal stem cells in human skin.
Aging rather than sun exposure is a major determining factor for the density of miR-125b-positive epidermal stem cells in human skin.
Pathology international (2015-06-18)
Shinya Toyokuni, Li Jiang, Shenqi Wang, Ayaka Hirao, Tamae Wada, Chieko Soh, Kazumi Toyama, Akira Kawada
ZUSAMMENFASSUNG
Sunlight exposure and aging are two major factors in the deterioration of skin function. In the present study, we used eighty formalin-fixed human skin samples from sun-exposed and unexposed areas from old and young individuals to evaluate the presence of miR-125b-positive epidermal stem cells (ESCs) by in situ hybridization. miR-125b-positive ESCs were detected in the basal layer of the epidermis. The density of miR-125b-positive ESCs was significantly associated with age rather than sun exposure, whereas the density of miR-125b-positive ESCs tended to decrease in the sun-exposed area. These data suggest the potential use of miR-125b as a surrogate marker for the quality of epidermal cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioUltra, Molecular Biology, 1 M in H2O