Merck
Lipoxin and resolvin biosynthesis is dependent on 5-lipoxygenase activating protein.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-08-21)
Christoph Lehmann, Julia Homann, Ann-Katrin Ball, René Blöcher, Thea K Kleinschmidt, Devaraj Basavarajappa, Carlo Angioni, Nerea Ferreirós, Ann-Kathrin Häfner, Olof Rådmark, Ewgenij Proschak, Jesper Z Haeggström, Gerd Geisslinger, Michael J Parnham, Dieter Steinhilber, Astrid Stefanie Kahnt
ZUSAMMENFASSUNG
Resolution of acute inflammation is an active process coordinated by proresolving lipid mediators (SPMs) such as lipoxins (LXs) and resolvins (Rvs), which are formed by the concerted action of 2 lipoxygenases (LOs). Because the exact molecular mechanisms of SPM biosynthesis are not completely understood, we aimed to investigate LX and D-type Rv formation in human leukocytes and HEK293T cells overexpressing leukotriene (LT) pathway enzymes. Activity assays in precursor (15-hydroxyeicosatetraenoic acids, 17-HDoHE)-treated granulocytes [polymorphonuclear leukocytes (PMNLs)] showed a strict dependence of LXA4/RvD1 biosynthesis on cell integrity, and incubation with recombinant human 5-LO did not lead to LX or Rv formation. Pharmacologic inhibition of 5-LO activating protein (FLAP) by MK-886 inhibited LXA4/RvD1 biosynthesis in precursor-treated PMNLs (drug concentration causing 50% inhibition ∼ 0.3/0.2 µM), as did knockdown of the enzyme in MM6 cells, and precursor-treated HEK293T overexpressing 5-LO produced high amounts of LXA4 only in the presence of FLAP. In addition, inhibition of cytosolic phospholipase A2α (cPLA2α) interfered with LXA4/RvD1 formation from exogenous precursors in PMNLs. Furthermore, inhibition of the LT synthases LTA4 hydrolase and LTC4 synthase in PMNL/platelet coincubations augmented LXA4 levels. These findings show that several enzymes known to be involved in the biosynthesis of proinflammatory LTs, such as FLAP and cPLA2α, also contribute to LX and Rv formation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
SAFC
L-Glutamin
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Oxalessigsäure, ≥97% (HPLC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Oxalessigsäure, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97% (HPLC)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Oxalessigsäure, powder, suitable for hybridoma
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.