Merck
  • Bioanalysis and enantioseparation of dl-carnitine in human plasma by the derivatization approach.
Bioanalysis and enantioseparation of dl-carnitine in human plasma by the derivatization approach.
Bioanalysis (2015-10-13)
Vinod Kumar Vashistha, Ravi Bhushan
ZUSAMMENFASSUNG
L-carnitine is an over the counter drug, used to treat disorders like cardiomyopathy, skeletal myopathy, hypoglycemia and hyperammonemia. Preparations containing D-carnitine should be avoided by dialysis patients because it has toxic influence on biochemical processes by inhibiting the carnitine acetyltransferase. Therefore, it is of utmost importance to assess and control the content of D-carnitine. A HPLC method was developed and validated for determination and enantiomeric resolution of DL-carnitine in human plasma by derivatization approach. (S)-Naproxen-based three derivatizing reagents were synthesized and applied. The limit of detection values were found to be 1.26 and 1.35 ng ml(-1) for the two isomers. The method is simple, reproducible, and can be used for routine analysis in laboratories for control of enantiomeric purity of carnitine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Borsäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
4-(Dimethylamino)-pyridin, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Borsäure, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Salzsäure, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid, 99%
Sigma-Aldrich
Borsäure, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Borsäure, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether