Merck
  • Molecular mechanisms of human thyrocyte dysfunction induced by low concentrations of polychlorinated biphenyl 118 through the Akt/FoxO3a/NIS pathway.
Molecular mechanisms of human thyrocyte dysfunction induced by low concentrations of polychlorinated biphenyl 118 through the Akt/FoxO3a/NIS pathway.
Journal of applied toxicology : JAT (2015-02-04)
Hongwei Guo, Hui Yang, Huanhuan Chen, Wen Li, Jinmei Tang, Pei Cheng, Yuchun Xie, Yun Liu, Guoxian Ding, Dai Cui, Xuqin Zheng, Yu Duan
ZUSAMMENFASSUNG
Polychlorinated biphenyls (PCBs) are typical persistent organic pollutants that can interfere with multiple organ systems of humans. Previously, we concluded that persistent exposure to low doses of PCB118 could severely damage the thyroidal structure, dramatically decrease the concentration of serum thyroid hormones and inhibit the pivotal gene expressions such as sodium/iodide symporter (NIS) and thyroglobulin (Tg). To explore the molecular mechanisms of thyrocyte dysfunction induced by 2,3',4,4',5-pentachlorobiphenyl (PCB118), monolayer cultured human thyroid epithelial cells (HTECs) were treated with PCB118 or dimethyl sulfoxide (DMSO) as a control. Our results indicated that relatively higher concentrations of PCB118 could induce a loss in the viability of HTEC. In cultures with concentrations of PCB118 from 0.025 to 25 nM, which did not affect cell viability or apoptosis, concentrations of Tg and thyroxine (T(4)) were significantly decreased compared with those in the controls. In addition, mRNA and protein levels of Akt were increased significantly in the PCB118-treated groups, whereas FoxO3a expression did not show particular variation. Furthermore, exposure to PCB118 was associated with a significant increase of the protein levels of p-Akt and p-FoxO3a, and these effects were blocked by LY294002. In contrast, mRNA and protein expression levels of NIS were decreased significantly, and this effect was blocked by LY294002. Unlike control cells, a cytoplasmic shift of FoxO3a was observed in the PCB118-treated group. Our research suggests that PCB118 may induce thyrocyte dysfunction through the Akt/FoxO3a/NIS signalling pathway, which provides potential new insights for finding interventions to counteract the damage to the human body caused by PCBs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Sigma-Aldrich
N-Iodsuccinimid, 95%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin-Fragment 26-33 Amid, ≥97% (HPLC), powder
Sigma-Aldrich
Hydrocortison, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®