Merck
Evidence for downregulation of the negative regulator SPRED2 in clinical prostate cancer.
British journal of cancer (2012-11-22)
N Kachroo, T Valencia, A Y Warren, V J Gnanapragasam
ZUSAMMENFASSUNG
SPRED1 and 2 are key negative regulators of MAPK signalling in mammalian cells. Here, we investigate the expression and functional role of SPREDs in prostate cancer. A transcriptome bank of microdissected grade-specific primary cancers was constructed and interrogated for transcript expression of prostate cancer genes, known negative signalling regulators as well as SPRED1 and 2. The effect of SPRED2 manipulation was tested in in vitro assays. In a panel of 5 benign glands and 15 tumours, we observed concomitant downregulation of the negative regulators SEF and DUSP1 in tumours with increasing Gleason grade. Profiling in the same cohorts revealed downregulation of SPRED2 mRNA in tumours compared with benign glands (P<0.05). By contrast, SPRED1 expression remained unchanged. This observation was further validated in two additional separate cohorts of microdissected tumours (total of n=10 benign and n=58 tumours) with specific downregulation of SPRED2 particularly in higher grade tumours. In functional assays, SPRED2 overexpression reduced ERK phosphorylation and inhibited prostate cancer cell proliferation and migration in response to different growth factors and full-media stimulation (P<0.001). Conversely, SPRED2 suppression by siRNA enhanced the mitogenic response to growth factors and full media (P<0.001). These data suggest first evidence that SPRED2 is downregulated in prostate cancer and warrants further investigation as a potential tumour-suppressor gene.