Merck
  • Porcine induced pluripotent stem cells require LIF and maintain their developmental potential in early stage of embryos.
Porcine induced pluripotent stem cells require LIF and maintain their developmental potential in early stage of embryos.
PloS one (2012-12-20)
De Cheng, Yanjie Guo, Zhenzhen Li, Yajun Liu, Xing Gao, Yi Gao, Xiang Cheng, Junhe Hu, Huayan Wang
ZUSAMMENFASSUNG
Porcine induced pluripotent stem (piPS) cell lines have been generated recently by using a cocktail of defined transcription factors, however, the features of authentic piPS cells have not been agreed upon and most of published iPS clones did not meet the stringent requirements of pluripotency. Here, we report the generation of piPS cells from fibroblasts using retrovirus carrying four mouse transcription factors (mOct4, mSox2, mKlf4 and mc-Myc, 4F). Multiple LIF-dependent piPS cell lines were generated and these cells showed the morphology similar to mouse embryonic stem cells and other pluripotent stem cells. In addition to the routine characterization, piPS cells were injected into porcine pre-compacted embryos to generate chimera embryos and nuclear transfer (NT) embryos. The results showed that piPS cells retain the ability to integrate into inner and outer layers of the blastocysts, and support the NT embryos development to blastocysts. The generations of chimera embryos and NT embryos derived from piPS clones are a practical means to determine the quality of iPS cells ex vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6-(Dimethylamino)purin, ≥98%
Millipore
JAK-Inhibitor I, JAK Inhibitor I, CAS 457081-03-7, is a potent, reversible, cell-permeable, and ATP-competitive inhibitor of JAK 1 (IC50 = 15 nM), JAK2 (IC50 = 1 nM), JAK3 (Ki = 5 nM) and Tyk2 (IC50 = 1 nM).