Merck
  • Aptamer-polymer functionalized silicon nanosubstrates for enhanced recovered circulating tumor cell viability and in vitro chemosensitivity testing.
Aptamer-polymer functionalized silicon nanosubstrates for enhanced recovered circulating tumor cell viability and in vitro chemosensitivity testing.
International journal of nanomedicine (2016-06-09)
Qinglin Shen, Caixia Peng, Yan Zhan, Liang Fan, Mengyi Wang, Qing Zhou, Jue Liu, Xiaojuan Lv, Qiu Tang, Jun Li, Xiaodong Huang, Jiahong Xia
ZUSAMMENFASSUNG
Selection of the optimal chemotherapy regimen for an individual cancer patient is challenging. The existing chemosensitivity tests are costly, time-consuming, and not amenable to wide utilization within a clinic. This limitation might be addressed by the recently proposed use of circulating tumor cells (CTCs), which provide an opportunity to noninvasively monitor response to therapy. Over the past few decades, various techniques were developed to capture and recover CTCs, but these techniques were often limited by a capture and recovery performance tradeoff between high viability and high efficiency. In this work, we used anti-epithelial cell adhesion molecule coated aptamer-poly (N-isopropylacrylamide) functionalized silicon nanowire substrates to capture and release epithelial cell adhesion molecule-positive CTCs at 32°C and 4°C, respectively. Then, we applied the nuclease to digest the aptamer to release the captured CTCs (near or at the end of the polymer brush), which cannot be released by heating/cooling process. High viability and purity CTCs could be achieved by decreasing the heating/cooling cycles and enzymatic treatment rounds. Furthermore, the time-saving process is helpful to maintain the morphology and enhance vitality of the recovered CTCs and is beneficial to the subsequent cell culture in vitro. We validated the feasibility of chemosensitivity testing based on the recovered HCC827 cells using an adenosine triphosphate-tumor chemosensitivity assay, and the results suggested that our method can determine which agent and what concentration have the best chemosensitivity for the culturing recovered CTCs. So, the novel method capable of a highly effective capture and recovery of high viability CTCs will pave the way for chemosensitivity testing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
GenomePlex® GGA Kit zur Gesamtgenom-Amplifikation, Optimized kit with enzyme for amplifying a variety of DNA including FFPE tissue