Merck
  • Proteomic analysis of the vitamin C effect on the doxorubicin cytotoxicity in the MCF-7 breast cancer cell line.
Proteomic analysis of the vitamin C effect on the doxorubicin cytotoxicity in the MCF-7 breast cancer cell line.
Journal of cancer research and clinical oncology (2016-09-14)
Peter Bober, Michal Alexovic, Ivan Talian, Zuzana Tomkova, Zuzana Viscorova, Maria Benckova, Igor Andrasina, Rachele Ciccocioppo, Daniel Petrovic, Mariusz Adamek, Peter Kruzliak, Jan Sabo
ZUSAMMENFASSUNG
Doxorubicin is an anthracycline drug which inhibits the growth of breast cancer cell lines. However, a major factor limiting its use is a cumulative, dose-dependent cardiotoxicity, resulting in a permanent loss of cardiomyocytes. Vitamin C was found to potentiate the cytotoxic effects of a variety of chemotherapeutic drugs including doxorubicin. The aim of the study was to describe the changes in protein expression and proliferation of the MCF-7 cells induced by the vitamin C applied with doxorubicin. Label-free quantitative proteomics and real-time cell analysis methods were used to search for proteome and cell proliferation changes. These changes were induced by the pure DOX and by DOX combined with vitamin C applied on the MCF-7 cell line. From the real-time cell analysis experiments, it is clear that the highest anti-proliferative effect occurs with the addition of 200 µM of vitamin C to 1 µM of doxorubicin. By applying both the label-free protein quantification method and total ion current assay, we found statistically significant changes (p ≤ 0.05) of 26 proteins induced by the addition of vitamin C to doxorubicin on the MCF-7 cell line. These differentially expressed proteins are involved in processes such as structural molecule activity, transcription and translation, immune system process and antioxidant, cellular signalling and transport. The detected proteins may be capable of predicting response to DOX therapy. This is a key tool in the treatment of breast cancer, and the combination with vit C seems to be of particular interest due to the fact that it can potentiate anti-proliferative effect of DOX.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-(−)-Glukose, ≥99%