- Exploring encapsulation mechanism of DNA and mononucleotides in sol-gel derived silica.
Exploring encapsulation mechanism of DNA and mononucleotides in sol-gel derived silica.
Journal of biomaterials applications (2017-06-02)
Derya Kapusuz, Caner Durucan
PMID28566001
ZUSAMMENFASSUNG
The encapsulation mechanism of DNA in sol-gel derived silica has been explored in order to elucidate the effect of DNA conformation on encapsulation and to identify the nature of chemical/physical interaction of DNA with silica during and after sol-gel transition. In this respect, double stranded DNA and dAMP (2'-deoxyadenosine 5'-monophosphate) were encapsulated in silica using an alkoxide-based sol-gel route. Biomolecule-encapsulating gels have been characterized using UV-Vis,