- Purification and characterization of an extracellular glucoamylase from the yeast Candida tsukubaensis CBS 6389.
Purification and characterization of an extracellular glucoamylase from the yeast Candida tsukubaensis CBS 6389.
Antonie van Leeuwenhoek (1985-01-01)
R De Mot, E Van Oudendijck, H Verachtert
PMID3937490
ZUSAMMENFASSUNG
The starch-degrading yeast Candida tsukubaensis CBS 6389 secreted amylase at high activity when grown in a medium containing soluble starch. The extracellular alpha-amylase activity was very low. The major amylase component was purified by DEAE-Sephadex A-50 chromatography and Ultrogel AcA 44 gel filtration and characterized as a glucoamylase. The enzyme proved to be a glycoprotein with a molecular weight of 56 000. The glucoamylase had a temperature optimum at 55 degrees C and displayed highest activity in a pH range of 2.4-4.8. Acarbose strongly inhibited the purified glucoamylase. Debranching activity was present as demonstrated by the release of glucose from pullulan.