Merck
  • Zinc oxide nanoparticles affect the expression of p53, Ras p21 and JNKs: an ex vivo/in vitro exposure study in respiratory disease patients.
Zinc oxide nanoparticles affect the expression of p53, Ras p21 and JNKs: an ex vivo/in vitro exposure study in respiratory disease patients.
Mutagenesis (2014-11-09)
Ashutosh Kumar, Mojgan Najafzadeh, Badie K Jacob, Alok Dhawan, Diana Anderson
ABSTRACT
Zinc oxide (ZnO) nanoparticles are the mostly used engineered metal oxide nanoparticles in consumer products. This has increased the likelihood of human exposure to this engineered nanoparticle (ENPs) through different routes. At present, the majority of the studies concerning ZnO ENPs toxicity have been conducted using in vitro and in vivo systems. In this study, for the first time we assessed the effect of ZnO ENPs on the major cellular pathways in the lymphocytes of healthy individuals as well as in susceptible patients suffering from lung cancer, chronic obstructive pulmonary disease (COPD) and asthma. Using the differential expression analysis, we observed a significant (P < 0.05) dose-dependent (10, 20 and 40 µg/ml for 6h) increase in the expression of tumour suppressor protein p53 (40, 60 and 110%); Ras p21 (30, 52 and 80%); c-Jun N-terminal kinases; JNKs) (28, 47 and 78%) in lung cancer patient samples treated with ZnO ENPs compared to healthy controls. A similar trend was also seen in COPD patient samples where a significant (P < 0.05) dose-dependent increase in the expression of tumour suppressor protein p53 (26, 45 and 84%), Ras p21 (21, 40 and 77%), JNKs (17, 32 and 69%) was observed after 6h of ZnO ENPs treatment at the aforesaid concentrations. However, the increase in the expression profile of tested protein was not significant in the asthma patients as compared to controls. Our results reiterate the concern about the safety of ZnO ENPs in consumer products and suggest the need for a complete risk assessment of any new ENPs before its use.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O