Merck
  • Regorafenib inhibits colorectal tumor growth through PUMA-mediated apoptosis.
Regorafenib inhibits colorectal tumor growth through PUMA-mediated apoptosis.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-04-26)
Dongshi Chen, Liang Wei, Jian Yu, Lin Zhang
ABSTRACT
Regorafenib, a multikinase inhibitor targeting the Ras/Raf/MEK/ERK pathway, has recently been approved for the treatment of metastatic colorectal cancer. However, the mechanisms of action of regorafenib in colorectal cancer cells have been unclear. We investigated how regorafenib suppresses colorectal cancer cell growth and potentiates effects of other chemotherapeutic drugs. We determined whether and how regorafenib induces the expression of PUMA, a p53 target and a critical mediator of apoptosis in colorectal cancer cells. We also investigated whether PUMA is necessary for the killing and chemosensitization effects of regorafenib in colorectal cancer cells. Furthermore, xenograft tumors were used to test if PUMA mediates the in vivo antitumor, antiangiogenic, and chemosensitization effects of regorafenib. We found that regorafenib treatment induces PUMA in colorectal cancer cells irrespective of p53 status through the NF-κB pathway following ERK inhibition and glycogen synthase kinase 3β activation. Upregulation of PUMA is correlated with apoptosis induction in different colorectal cancer cell lines. PUMA is necessary for regorafenib-induced apoptosis in colorectal cancer cells. Chemosensitization by regorafenib is mediated by enhanced PUMA induction through different pathways. Furthermore, deficiency in PUMA abrogates the in vivo antitumor, antiangiogenic, and chemosensitization effects of regorafenib. Our results demonstrate a key role of PUMA in mediating the anticancer effects of regorafenib in colorectal cancer cells. They suggest that PUMA induction can be used as an indicator of regorafenib sensitivity, and also provide a rationale for manipulating the apoptotic machinery to improve the therapeutic efficacy of regorafenib and other targeted drugs.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
MISSION® esiRNA, targeting human KDR
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Kdr
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%