Merck
  • Improving vagal activity ameliorates cardiac fibrosis induced by angiotensin II: in vivo and in vitro.
Improving vagal activity ameliorates cardiac fibrosis induced by angiotensin II: in vivo and in vitro.
Scientific reports (2015-11-26)
Jin-Jun Liu, Ning Huang, Yi Lu, Mei Zhao, Xiao-Jiang Yu, Yang Yang, Yong-hua Yang, Wei-Jin Zang
ABSTRACT
Cardiac remodeling is characterized by overactivity of the renin-angiotensin system (RAS) and withdrawal of vagal activity. We hypothesized that improving vagal activity could attenuate cardiac fibrosis induced by angiotensin II (Ang II) in vivo and in vitro. Rats were subjected to abdominal aorta constriction (AAC) with or without pyridostigmine (PYR) (31 mg/kg/d). After 8 weeks, PYR significantly decreased Ang II level, AT1 protein expression, and collagen deposition in cardiac tissue and improved heart rate variability, baroreflex sensitivity and cardiac function, which were abolished by atropine. In vitro, treatment of cardiac fibroblasts (CFs) with Ang II (10(-7) M) increased cell proliferation, migration, transformation, and secretory properties, which were significantly diminished by acetylcholine (ACh, 10(-6) M). Subsequently, Ang II significantly increased collagen type I expression as well as metalloproteinase (MMP)-2 expression and activity. Transforming growth factor (TGF)-β1 expression and Smad3 phosphorylation presented a similar trend. Notably, the knockdown of the acetylcholine M2 receptor by siRNA could abolish ACh anti-fibrotic action. These data implicated cholinesterase inhibitor can increase vagal activity and reduce local Ang II level, and ACh inhibit Ang II pro-fibrotic effects. Our findings suggested that the parasympathetic nervous system can serve as a promising target for cardiac remodeling treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human CHRM2
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rasl2-9-Ps
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%