Vai a
TMO2425-1003
Nalgene® narrow-mouth Right-to-Know wash bottle
low-density polyethylene, bottle volume 1000 mL, methanol, case of 12 ea
Sinonimo/i:
Nalgene bottle, plastic bottle, Nalgene 2425-1003, dispensing bottle, fluid dispenser, wash bottle
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Voi/SKUDisponibilitàPrezzo
12 ea
Per conoscere la disponibilità, visualizza il carrello
CHF 242.00
Informazioni su questo articolo
NACRES:
NB.43
UNSPSC Code:
41121800
eCl@ss:
32049090
CHF 242.00
Per conoscere la disponibilità, visualizza il carrello
​
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
low-density polyethylene , narrow-mouth
packaging
case of 12 ea
manufacturer/tradename
Nalgene 2425-1003
bottle volume
1000 mL
General description
Meet regulatory labeling requirements for chemical containers with Thermo Scientific Nalgene Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable symbols and colors. These low density polyethylene bottles are flexible for easy dispensing and translucent for viewing contents.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
TMO24251002TMO24251004TMO24251005
description
low-density polyethylene, bottle volume 1000 mL, methanol, case of 12 ea
description
low-density polyethylene, bottle volume 1000 mL, ethyl alcohol, case of 12 ea
description
low-density polyethylene, bottle volume 1000 mL, isopropanol, case of 12 ea
description
low-density polyethylene, bottle volume 1000 mL, distilled water, case of 12 ea
manufacturer/tradename
Nalgene 2425-1003
manufacturer/tradename
Nalgene 2425-1002
manufacturer/tradename
Nalgene 2425-1004
manufacturer/tradename
Nalgene 2425-1005
material
narrow-mouth , low-density polyethylene
material
narrow-mouth , low-density polyethylene
material
low-density polyethylene , narrow-mouth
material
low-density polyethylene , narrow-mouth
packaging
case of 12 ea
packaging
case of 12 ea
packaging
case of 12 ea
packaging
case of 12 ea
bottle volume
1000 mL
bottle volume
1000 mL
bottle volume
1000 mL
bottle volume
1000 mL
Still not finding the right product?
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)